man

Test

CAS RN 1013640-87-3

BNHNFFFK

CAS RN 1013640-87-3

IUPAC Name: potassium trifluoro(1H-pyrazol-5-yl)boranuide
Synonyms: potassium trifluoro(2H-pyrazol-3-yl)boranuide
Molecular Weight (g/mol): 173.97
Molecular Formula: C3H3BF3KN2
InChi Key: QNHYBMZHGRWURK-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results