accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH4FeOSOOOOSOOO

CAS RN 10138-04-2

IUPAC Name: iron(3+) ammonium disulfate
Synonyms: ammonium iron(III) sulfate
Molecular Weight (g/mol): 266
Molecular Formula: FeH4NO8S2
InChi Key: XGGLLRJQCZROSE-UHFFFAOYSA-K
SMILES: [NH4+].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results