accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1013916-37-4

IUPAC Name: 2-chloro-8-cyclopentyl-5-methyl-7H,8H-pyrido[2,3-d]pyrimidin-7-one
Synonyms: 2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7-one
Molecular Weight (g/mol): 263.73
Molecular Formula: C13H14ClN3O
InChi Key: BSKNQSYIDZUXQT-UHFFFAOYSA-N
SMILES: CC1=CC(=O)N(C2CCCC2)C2=NC(Cl)=NC=C12
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results