accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 101463-69-8

IUPAC Name: 3-{4-[2-chloro-4-(trifluoromethyl)phenoxy]-2-fluorophenyl}-1-(2,6-difluorobenzoyl)urea
Synonyms: flufenoxuron cascade
Molecular Weight (g/mol): 488.77
Molecular Formula: C21H11ClF6N2O3
InChi Key: RYLHNOVXKPXDIP-UHFFFAOYSA-N
SMILES: FC1=CC(OC2=CC=C(C=C2Cl)C(F)(F)F)=CC=C1NC(=O)NC(=O)C1=C(F)C=CC=C1F
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results