accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1016-58-6

IUPAC Name: (4,5-dimethoxy-2-nitrophenyl)methanol
Synonyms: (4,5-dimethoxy-2-nitrophenyl)methanol
Molecular Weight (g/mol): 213.19
Molecular Formula: C9H11NO5
InChi Key: WBSCOJBVYHQOFB-UHFFFAOYSA-N
SMILES: COC1=CC(CO)=C(C=C1OC)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results