accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 101664-56-6

ONONOClF

CAS RN 101664-56-6

IUPAC Name: 2-chloro-3-fluoro-4-nitropyridin-1-ium-1-olate
Synonyms: 2-chloro-3-fluoro-4-nitropyridin-1-ium-1-olate
Molecular Weight (g/mol): 192.53
Molecular Formula: C5H2ClFN2O3
InChi Key: IQKSCEDCDQGAAO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=[N+]([O-])C(Cl)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results