accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NSNN

CAS RN 1016888-78-0

IUPAC Name: 2-[4-(pyridin-4-yl)-1,3-thiazol-2-yl]acetonitrile
Synonyms: 2-[4-(pyridin-4-yl)-1,3-thiazol-2-yl]acetonitrile
Molecular Weight (g/mol): 201.25
Molecular Formula: C10H7N3S
InChi Key: LIIDMCZQVRSSQE-UHFFFAOYSA-N
SMILES: N#CCC1=NC(=CS1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results