CAS RN 1017778-34-5
CAS RN 1017778-34-5
IUPAC Name:
2-fluoro-4,6-bis(trifluoromethyl)benzoyl chloride
Synonyms:
2-fluoro-4,6-bis(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol):
294.55
Molecular Formula:
C9H2ClF7O
InChi Key:
RBJVFNYOFYHETI-UHFFFAOYSA-N
SMILES:
FC1=CC(=CC(=C1C(Cl)=O)C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™
CAS: 1017778-34-5 Molecular Formula: C9H2ClF7O Molecular Weight (g/mol): 294.553 MDL Number: MFCD09258670 InChI Key: RBJVFNYOFYHETI-UHFFFAOYSA-N Synonym: 2-fluoro-4,6-bis trifluoromethyl benzoyl chloride PubChem CID: 46737582 IUPAC Name: 2-fluoro-4,6-bis(trifluoromethyl)benzoyl chloride SMILES: C1=C(C=C(C(=C1C(F)(F)F)C(=O)Cl)F)C(F)(F)F