CAS RN 1017778-52-7
CAS RN 1017778-52-7
IUPAC Name:
3-chloro-5-(trifluoromethoxy)phenol
Synonyms:
3-chloro-5-(trifluoromethoxy)phenol
Molecular Weight (g/mol):
212.55
Molecular Formula:
C7H4ClF3O2
InChi Key:
GDQVMGRMDBOXLX-UHFFFAOYSA-N
SMILES:
OC1=CC(OC(F)(F)F)=CC(Cl)=C1
1
–
1
of
1
results
1
3-Chloro-5-(trifluoromethoxy)phenol, 97%, Thermo Scientific™
CAS: 1017778-52-7 Molecular Formula: C7H4ClF3O2 Molecular Weight (g/mol): 212.55 MDL Number: MFCD09832330 InChI Key: GDQVMGRMDBOXLX-UHFFFAOYSA-N Synonym: 3-chloro-5-trifluoromethoxy phenol,5-chloro-3-trifluoromethoxy phenol,3-chloro-5-trifluoromethoxy phenol, jrd PubChem CID: 46737653 IUPAC Name: 3-chloro-5-(trifluoromethoxy)phenol SMILES: OC1=CC(OC(F)(F)F)=CC(Cl)=C1