CAS RN 1017778-95-8
CAS RN 1017778-95-8
IUPAC Name:
1-[2-methoxy-6-(trifluoromethyl)phenyl]methanamine
Synonyms:
1-[2-methoxy-6-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol):
205.18
Molecular Formula:
C9H10F3NO
InChi Key:
CPYXGRVUWLCIHG-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(=C1CN)C(F)(F)F
1
–
1
of
1
results
1
2-Methoxy-6-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™
CAS: 1017778-95-8 Molecular Formula: C9H10F3NO Molecular Weight (g/mol): 205.18 MDL Number: MFCD09832323 InChI Key: CPYXGRVUWLCIHG-UHFFFAOYSA-N Synonym: 2-methoxy-6-trifluoromethyl benzylamine,1-2-methoxy-6-trifluoromethyl phenyl methanamine,2-methoxy-6-trifluoromethyl benzylamine, jrd,2-methoxy-6-trifluoromethyl phenyl methanamine,6-methoxy-2-trifluoromethyl phenyl methylamine PubChem CID: 46737594 SMILES: COC1=CC=CC(=C1CN)C(F)(F)F