accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1017779-36-0

OFFBrF

CAS RN 1017779-36-0

IUPAC Name: 2-(bromomethyl)-4-(difluoromethoxy)-1-fluorobenzene
Synonyms: 2-(bromomethyl)-4-(difluoromethoxy)-1-fluorobenzene
Molecular Weight (g/mol): 255.03
Molecular Formula: C8H6BrF3O
InChi Key: IYQMYENSWWJQAE-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC(CBr)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results