accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1017779-54-2

OFFBrCl

CAS RN 1017779-54-2

IUPAC Name: 2-(bromomethyl)-1-chloro-4-(difluoromethoxy)benzene
Synonyms: 2-(bromomethyl)-1-chloro-4-(difluoromethoxy)benzene
Molecular Weight (g/mol): 271.49
Molecular Formula: C8H6BrClF2O
InChi Key: NLCSHYHRPBCBDK-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC(CBr)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results