accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1017781-60-0

ONNSBr

CAS RN 1017781-60-0

IUPAC Name: 4-(4-bromo-1,3-thiazol-2-yl)morpholine
Synonyms: 4-(4-bromo-1,3-thiazol-2-yl)morpholine
Molecular Weight (g/mol): 249.13
Molecular Formula: C7H9BrN2OS
InChi Key: XPFAGXZWVZAYCS-UHFFFAOYSA-N
SMILES: BrC1=CSC(=N1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results