accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1019540-88-5

ClHNF

CAS RN 1019540-88-5

IUPAC Name: 3-chloro-N-[(4-fluorophenyl)methyl]aniline
Synonyms: 3-chloro-N-[(4-fluorophenyl)methyl]aniline
Molecular Weight (g/mol): 235.69
Molecular Formula: C13H11ClFN
InChi Key: IAALBLSPXNWMLD-UHFFFAOYSA-N
SMILES: FC1=CC=C(CNC2=CC(Cl)=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results