accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 102-25-0

H3CCH3H3C

CAS RN 102-25-0

IUPAC Name: 1,3,5-triethylbenzene
Synonyms: 1,3,5-triethylbenzene
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: WJYMPXJVHNDZHD-UHFFFAOYSA-N
SMILES: CCC1=CC(CC)=CC(CC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results