accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 102-87-4

CH3NCH3CH3

CAS RN 102-87-4

IUPAC Name: tridodecylamine
Synonyms: trilaurylamine
Molecular Weight (g/mol): 522
Molecular Formula: C36H75N
InChi Key: SWZDQOUHBYYPJD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCN(CCCCCCCCCCCC)CCCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results