accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 102-97-6

IUPAC Name: benzyl(propan-2-yl)amine
Synonyms: isopropylbenzylamine
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: LYBKPDDZTNUNNM-UHFFFAOYSA-N
SMILES: CC(C)NCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)