accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1021298-67-8

IUPAC Name: 1-cyclopropanecarbonylpiperazine hydrochloride
Synonyms: 1-cyclopropanecarbonylpiperazine hydrochloride
Molecular Weight (g/mol): 190.67
Molecular Formula: C8H15ClN2O
InChi Key: WIHDAPMHNYYTOA-UHFFFAOYSA-N
SMILES: Cl.O=C(C1CC1)N1CCNCC1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results