accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10214-39-8

BOOBOOH2OPb

CAS RN 10214-39-8

IUPAC Name: λ²-lead(2+) hydrate dioxoborinate
Synonyms: λ²-lead(2+) hydrate dioxoborinate
Molecular Weight (g/mol): 310.8
Molecular Formula: B2H2O5Pb
InChi Key: RTSNZWSJUUUHCZ-UHFFFAOYSA-N
SMILES: O.[Pb++].[O-]B=O.[O-]B=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results