accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 102191-92-4

CH3CH3H3CSiCH3H3COO

CAS RN 102191-92-4

IUPAC Name: 2-[(tert-butyldimethylsilyl)oxy]acetaldehyde
Synonyms: 2-[(tert-butyldimethylsilyl)oxy]acetaldehyde
Molecular Weight (g/mol): 174.32
Molecular Formula: C8H18O2Si
InChi Key: MEBFFOKESLAUSJ-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OCC=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)