accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1022-13-5

IUPAC Name: 2-benzoyl-4-chloro-N-methylaniline
Synonyms: 2-benzoyl-4-chloro-N-methylaniline
Molecular Weight (g/mol): 245.71
Molecular Formula: C14H12ClNO
InChi Key: WPNMLCMTDCANOZ-UHFFFAOYSA-N
SMILES: CNC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results