accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNNH2

CAS RN 102308-97-4

IUPAC Name: 1-methyl-1H-indol-5-amine
Synonyms: 1-methylindol-5-amine
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: PGTSGPCXPIFQEL-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC(N)=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)