accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COClOHNSOONHONH

CAS RN 10238-21-8

IUPAC Name: 5-chloro-N-[2-(4-{[(cyclohexylcarbamoyl)amino]sulfonyl}phenyl)ethyl]-2-methoxybenzamide
Synonyms: glyburide glimide semi-euglucon abbenclamide gilemal glibenclamid heumann glibenclamid-ratiopharm glycolande renabetic semi-daonil
Molecular Weight (g/mol): 494
Molecular Formula: C23H28ClN3O5S
InChi Key: ZNNLBTZKUZBEKO-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C=C1C(=O)NCCC1=CC=C(C=C1)S(=O)(=O)NC(=O)NC1CCCCC1
Molecular Weight (g/mol) 
  • (7)
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (5)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Certifications/Compliance 
  • (3)
  • (1)
Industry Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (5)
  • (7)
  • (5)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)