accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 102392-87-0

IUPAC Name: morpholine-4-carboximidamide hydroiodide
Synonyms: morpholine-4-carboximidamide hydroiodide
Molecular Weight (g/mol): 257.08
Molecular Formula: C5H12IN3O
InChi Key: LJQSGLRGQQZIMF-UHFFFAOYSA-N
SMILES: I.NC(=N)N1CCOCC1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results