accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrNNClCl

CAS RN 102393-82-8

IUPAC Name: 6-bromo-2,4-dichloroquinazoline
Synonyms: 6-bromo-2,4-dichloroquinazoline
Molecular Weight (g/mol): 277.93
Molecular Formula: C8H3BrCl2N2
InChi Key: LBAYOWRVZAKPLS-UHFFFAOYSA-N
SMILES: ClC1=NC2=CC=C(Br)C=C2C(Cl)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results