accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 102394-31-0

H3CNCH3NONOHNN

CAS RN 102394-31-0

IUPAC Name: 2-(2-{2-[(diethylamino)methyl]piperidin-1-yl}acetyl)-2,4,9-triazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Synonyms: 2-(2-{2-[(diethylamino)methyl]piperidin-1-yl}acetyl)-2,4,9-triazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Molecular Weight (g/mol): 421.55
Molecular Formula: C24H31N5O2
InChi Key: UBRKDAVQCKZSPO-UHFFFAOYNA-N
SMILES: CCN(CC)CC1CCCCN1CC(=O)N1C2=CC=CC=C2C(=O)NC2=CC=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results