accessibility menu, dialog, popup

UserName

CAS RN 1024-99-3

CAS RN 1024-99-3

IUPAC Name: 1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 370.1
Molecular Formula: C9H11IN2O6
InChi Key: RKSLVDIXBGWPIS-UHFFFAOYNA-N
SMILES: OCC1OC(C(O)C1O)N1C=C(I)C(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results