CAS RN 10242-00-9
IUPAC Name:
6-nitro-1H-indole-2-carboxylic acid
Synonyms:
6-nitro-1H-indole-2-carboxylic acid
Molecular Weight (g/mol):
206.16
Molecular Formula:
C9H6N2O4
InChi Key:
JIXJNWVLWQQNDB-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC2=CC=C(C=C2N1)[N+]([O-])=O
1
–
1
of
1
results
1
6-Nitroindole-2-carboxylic acid, 97%, Thermo Scientific Chemicals
CAS: 10242-00-9 Molecular Formula: C9H6N2O4 Molecular Weight (g/mol): 206.157 MDL Number: MFCD02664466 InChI Key: JIXJNWVLWQQNDB-UHFFFAOYSA-N Synonym: 6-nitroindole-2-carboxylic acid,1h-indole-2-carboxylic acid, 6-nitro,6-nitro-2-indolecarboxylic acid,1h-indole-2-carboxylicacid, 6-nitro,6-nitroindol-2-carboxylic acid PubChem CID: 4714997 IUPAC Name: 6-nitro-1H-indole-2-carboxylic acid SMILES: C1=CC2=C(C=C1[N+](=O)[O-])NC(=C2)C(=O)O