accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNO

CAS RN 102432-05-3

IUPAC Name: 3-[(1H-imidazol-1-yl)methyl]benzaldehyde
Synonyms: 3-(imidazol-1-ylmethyl)benzaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: JMKQKKQFLYPQDD-UHFFFAOYSA-N
SMILES: O=CC1=CC(CN2C=CN=C2)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results