accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1025113-70-5

BONOFFFK

CAS RN 1025113-70-5

IUPAC Name: potassium trifluoro[3-(morpholine-4-carbonyl)phenyl]boranuide
Synonyms: potassium trifluoro[3-(morpholine-4-carbonyl)phenyl]boranuide
Molecular Weight (g/mol): 297.13
Molecular Formula: C11H12BF3KNO2
InChi Key: VMMCKOVDXHKYLW-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC(=CC=C1)C(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results