CAS RN 10255-95-5
IUPAC Name:
2-phenylpropanediamide
Synonyms:
2-phenylpropanediamide
Molecular Weight (g/mol):
178.19
Molecular Formula:
C9H10N2O2
InChi Key:
CPSUAFUQJBJMPO-UHFFFAOYSA-N
SMILES:
NC(=O)C(C(N)=O)C1=CC=CC=C1