accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10257-55-3

CaOHSOHO

CAS RN 10257-55-3

IUPAC Name: calcium sulfurous acid
Synonyms: calcium sulfite
Molecular Weight (g/mol): 122.15
Molecular Formula: CaH2O3S
InChi Key: GBAOBIBJACZTNA-UHFFFAOYSA-N
SMILES: [Ca++].OS(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results