accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH2HOOOOH

CAS RN 102593-74-8

IUPAC Name: 4,6-dibenzoyl-2-(prop-2-en-1-yl)benzene-1,3-diol
Synonyms: 4,6-dibenzoyl-2-(prop-2-en-1-yl)benzene-1,3-diol
Molecular Weight (g/mol): 358.39
Molecular Formula: C23H18O4
InChi Key: FSYGSBMXRNPJAD-UHFFFAOYSA-N
SMILES: OC1=C(CC=C)C(O)=C(C=C1C(=O)C1=CC=CC=C1)C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results