CAS RN 1026016-84-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(-)-Tetrabenazine (>75% ee), TRC
CAS: 1026016-84-1 Molecular Formula: C19H27NO3 Molecular Weight (g/mol): 317.42 Synonym: (3S,11bS)-1,3,4,6,7,11b-Hexahydro-9,10-dimethoxy-3-(2-methylpropyl)-2H-benzo[a]quinolizin-2-one,(-)-TBZ,(-)-Tetrabenazine,(3S,11bS)-TBZ,(3S,11bS)-Tetrabenazine IUPAC Name: (3S,11bS)-9,10-dimethoxy-3-(2-methylpropyl)-1,3,4,6,7,11b-hexahydrobenzo[a]quinolizin-2-one SMILES: COc1cc2CCN3C[C@H](CC(C)C)C(=O)C[C@H]3c2cc1OC