accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 102696-71-9

SOOH(E/Z)

CAS RN 102696-71-9

IUPAC Name: 3-(thiophen-3-yl)prop-2-enoic acid
Synonyms: 3-(thiophen-3-yl)prop-2-enoic acid
Molecular Weight (g/mol): 154.18
Molecular Formula: C7H6O2S
InChi Key: VYRYYUKILKRGDN-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CSC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results