accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOFFFK

CAS RN 1027642-30-3

IUPAC Name: potassium trifluoro(phenoxymethyl)boranuide
Synonyms: potassium trifluoro(phenoxymethyl)boranuide
Molecular Weight (g/mol): 214.04
Molecular Formula: C7H7BF3KO
InChi Key: OVYSNXCRBZPJIG-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)COC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results