accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1028-33-7

H3CNONCH3NNH3CO

CAS RN 1028-33-7

IUPAC Name: 1-hexyl-3,7-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: pentifylline SK 7 (pharmaceutical) pentifyllin hexyltheobromine cosadon pentiphylline cosaldon pentifilina pentifyllinum
Molecular Weight (g/mol): 264.33
Molecular Formula: C13H20N4O2
InChi Key: MRWQRJMESRRJJB-UHFFFAOYSA-N
SMILES: CCCCCCN1C(=O)N(C)C2=C(N(C)C=N2)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)