accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SNNOH

CAS RN 1028352-86-4

IUPAC Name: 2-[4-(4-hydroxyphenyl)-1,3-thiazol-2-yl]acetonitrile
Synonyms: 2-[4-(4-hydroxyphenyl)-1,3-thiazol-2-yl]acetonitrile
Molecular Weight (g/mol): 216.26
Molecular Formula: C11H8N2OS
InChi Key: CMIGLFUTBISGQV-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CSC(CC#N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results