accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10294-64-1

IUPAC Name: dipotassium dioxomanganesebis(olate)
Synonyms: potassium manganate
Molecular Weight (g/mol): 197.13
Molecular Formula: K2MnO4
InChi Key: XKYFHUXZPRFUTH-UHFFFAOYSA-N
SMILES: [K+].[K+].[O-][Mn]([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results