accessibility menu, dialog, popup

UserName

CAS RN 1029684-37-4

CAS RN 1029684-37-4

IUPAC Name: 1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole
Synonyms: 1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)imidazole
Molecular Weight (g/mol): 278.16
Molecular Formula: C14H23BN2O3
InChi Key: BVIWGBKKGPHNAB-UHFFFAOYNA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=CN1C1CCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results