accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 103-29-7

IUPAC Name: (2-phenylethyl)benzene
Synonyms: dibenzyl dihydrostilbene dibenzil 1,2-dihydrostilbene bibenzyl
Molecular Weight (g/mol): 182.27
Molecular Formula: C14H14
InChi Key: QWUWMCYKGHVNAV-UHFFFAOYSA-N
SMILES: C(CC1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (7)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (4)
  • (4)