accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COO(E)

CAS RN 103-36-6

IUPAC Name: ethyl (2E)-3-phenylprop-2-enoate
Synonyms: ethyl cinnamate
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: KBEBGUQPQBELIU-CMDGGOBGSA-N
SMILES: CCOC(=O)\C=C\C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (5)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (4)