accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO(E)

CAS RN 103-41-3

IUPAC Name: benzyl (2E)-3-phenylprop-2-enoate
Synonyms: benzyl cinnamate
Molecular Weight (g/mol): 238.29
Molecular Formula: C16H14O2
InChi Key: NGHOLYJTSCBCGC-VAWYXSNFSA-N
SMILES: O=C(OCC1=CC=CC=C1)\C=C\C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (8)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (4)
  • (5)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (4)
  • (8)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)