CAS RN 103-43-5
IUPAC Name:
1,4-dibenzyl butanedioate
Synonyms:
dibenzyl succinate
Molecular Weight (g/mol):
298.34
Molecular Formula:
C18H18O4
InChi Key:
ODBOBZHTGBGYCK-UHFFFAOYSA-N
SMILES:
O=C(CCC(=O)OCC1=CC=CC=C1)OCC1=CC=CC=C1
1
–
1
of
1
results
1
Dibenzyl succinate, 98%
CAS: 103-43-5 Molecular Formula: C18H18O4 Molecular Weight (g/mol): 298.34 MDL Number: MFCD00022034 InChI Key: ODBOBZHTGBGYCK-UHFFFAOYSA-N Synonym: dibenzyl succinate,butanedioic acid, bis phenylmethyl ester,spasmine,benzyl succinate,dibenzylsuccinate,succinic acid dibenzyl ester,unii-ck14ug55av,ck14ug55av,diphenylmethyl butane-1,4-dioate,succinic acid, dibenzyl ester PubChem CID: 7653 ChEBI: CHEBI:3058 IUPAC Name: dibenzyl butanedioate SMILES: O=C(CCC(=O)OCC1=CC=CC=C1)OCC1=CC=CC=C1