accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 103-70-8

IUPAC Name: N-phenylformamide
Synonyms: formanilide
Molecular Weight (g/mol): 121.14
Molecular Formula: C7H7NO
InChi Key: DYDNPESBYVVLBO-UHFFFAOYSA-N
SMILES: O=CNC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (3)