accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClON

CAS RN 103140-12-1

IUPAC Name: 2-(3,5-dichlorophenoxy)acetonitrile
Synonyms: 2-(3,5-dichlorophenoxy)acetonitrile
Molecular Weight (g/mol): 202.03
Molecular Formula: C8H5Cl2NO
InChi Key: PNROREDTZJCOHF-UHFFFAOYSA-N
SMILES: ClC1=CC(OCC#N)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results