CAS RN 1032564-18-3
CAS RN 1032564-18-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)glycine, TRC
CAS: 1032564-18-3 Molecular Formula: C12H19NO3 Molecular Weight (g/mol): 225.28 Synonym: 3-Hydroxy-1-adamantyl-D-glycine,2-((3-Hydroxyadamantan-1-yl)amino)acetic Acid IUPAC Name: 2-[(3-hydroxy-1-adamantyl)amino]acetic acid SMILES: OC(=O)CNC12CC3CC(CC(O)(C3)C1)C2