CAS RN 10332-33-9
CAS RN 10332-33-9
IUPAC Name:
sodium hydrate [(oxoboranyl)oxy]olate
Synonyms:
sodium hydrate (oxoboranyl)oxyolate
Molecular Weight (g/mol):
99.81
Molecular Formula:
BH2NaO4
InChi Key:
MDGXUEVTGARGDK-UHFFFAOYSA-M
SMILES:
O.[Na+].[O-]OB=O
1
–
1
of
1
results
1
Sodium perborate monohydrate, 95%, Thermo Scientific Chemicals
CAS: 10332-33-9 Molecular Formula: BNaO3·H2O Molecular Weight (g/mol): 99.81 MDL Number: MFCD00149230