accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FOONHCNOO

CAS RN 103361-09-7

IUPAC Name: 2-[7-fluoro-3-oxo-4-(prop-2-yn-1-yl)-3,4-dihydro-2H-1,4-benzoxazin-6-yl]-2,3,4,5,6,7-hexahydro-1H-isoindole-1,3-dione
Synonyms: flumioxazin sumisoya
Molecular Weight (g/mol): 354.34
Molecular Formula: C19H15FN2O4
InChi Key: FOUWCSDKDDHKQP-UHFFFAOYSA-N
SMILES: FC1=CC2=C(C=C1N1C(=O)C3=C(CCCC3)C1=O)N(CC#C)C(=O)CO2
Molecular Weight (g/mol) 
  • (3)
  • (4)
Concentration 
  • (2)
  • (2)
Solvent or Matrix 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results