accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 103361-67-7

OHNOFNOO

CAS RN 103361-67-7

IUPAC Name: 7-fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 7-fluoro-6-nitro-2,4-dihydro-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 212.14
Molecular Formula: C8H5FN2O4
InChi Key: MNUBUFBVOBPANC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(F)C=C2OCC(=O)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results